Anti-SERINC2 Rabbit Polyclonal Antibody (Biotin)

Proveedor: United States Biological
USBI041569-BIOTINEA 852 EUR
USBI041569-BIOTIN
Anti-SERINC2 Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: SERINC2 (serine incorporator 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR