Anti-SERINC2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Proveedor: United States Biological
USBI041569-APCEA 852 EUR
USBI041569-APC
Anti-SERINC2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SERINC2 (serine incorporator 2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human