Anti-NCL Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Proveedor: US Biological
USBI038834-PEEA 852 EUR
USBI038834-PE
Anti-NCL Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: NCL (nucleolin)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human