Anti-NCL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Proveedor: US Biological
USBI038834-FITCEA 852 EUR
USBI038834-FITC
Anti-NCL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: NCL (nucleolin)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR