Anti-NCL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Proveedor: US Biological
USBI038833-FITCEA 852 EUR
USBI038833-FITC
Anti-NCL Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Immunofluorescence: 1:10-50
Do not freeze!
Type: Primary
Antigen: NCL (nucleolin)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human