Anti-NCKPL Rabbit Polyclonal Antibody (Biotin)
Proveedor: US Biological
USBI038832-BIOTINEA 852 EUR
USBI038832-BIOTIN
Anti-NCKPL Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: NCKPL
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse