Anti-NCKPL Rabbit Polyclonal Antibody

Proveedor: US Biological
USBI038832EA 587 EUR
USBI038832
Anti-NCKPL Rabbit Polyclonal Antibody
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: NCKPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR