Anti-NCKAP1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Proveedor: US Biological
USBI038831-APCEA 852 EUR
USBI038831-APC
Anti-NCKAP1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: NCKAP1 (NCK-associated protein 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human