Anti-NCK1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Proveedor: US Biological
USBI038829-APEA 852 EUR
USBI038829-AP
Anti-NCK1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: NCK1 (NCK adaptor protein 1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human