Anti-NCF1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Proveedor: US Biological
USBI038828-PEEA 852 EUR
USBI038828-PE
Anti-NCF1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticuerpos
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!

Type: Primary
Antigen: NCF1 (neutrophil cytosolic factor 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR