Anti-NCF1 Rabbit Polyclonal Antibody (Biotin)
Proveedor: US Biological
USBI038828-BIOTINEA 852 EUR
USBI038828-BIOTIN
Anti-NCF1 Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Type: Primary
Antigen: NCF1 (neutrophil cytosolic factor 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human