Anti-P2RY12 Rabbit Polyclonal Antibody
Proveedor: Abcam
ABCAAB140862-50EA 550 EUR
ABCAAB140862-50
Anti-P2RY12 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: P2RY12 (purinergic receptor P2Y, G-protein coupled, 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human