Anti-ARPC1A Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB133160-200EA 520 EUR
ABCAAB133160-200
Anti-ARPC1A Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: ARPC1A (actin related protein 2/3 complex, subunit 1A, 41kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR