Anti-AKT2 Rabbit Monoclonal Antibody [clone: EP1676]

Proveedor: Abcam
ABCAAB131168-100EA 880 EUR
ABCAAB131168-100
Anti-AKT2 Rabbit Monoclonal Antibody [clone: EP1676]
Anticuerpos
Type: Primary
Antigen: AKT2 (v-akt murine thymoma viral oncogene homolog 2)
Clonality: monoclonal
Clone: EP1676
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR