Anti-ALOX5AP Rabbit Monoclonal Antibody [clone: EPR5640]

Proveedor: Abcam
ABCAAB124714-100EA 535 EUR
ABCAAB124714-100
Anti-ALOX5AP Rabbit Monoclonal Antibody [clone: EPR5640]
Anticuerpos
Type: Primary
Antigen: ALOX5AP (arachidonate 5-lipoxygenase-activating protein)
Clonality: monoclonal
Clone: EPR5640
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR