Anti-PLA2G15 Rabbit Polyclonal Antibody
Proveedor: Abcam
ABCAAB111898-50EA 920 EUR
ABCAAB111898-50
Anti-PLA2G15 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PLA2G15 (phospholipase A2, group XV)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human