Anti-DIABLO Mouse monoclonal antibody unconjugated [clone: SMAC 17 1-87]
Proveedor: Abcam
ABCAAB111893-100EA 530 EUR
ABCAAB111893-100
Anti-DIABLO Mouse monoclonal antibody unconjugated [clone: SMAC 17 1-87]
Anticuerpos
Type: Primary
Antigen: DIABLO (diablo, IAP-binding mitochondrial protein)
Clonality: monoclonal
Clone: SMAC 17 1-87
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human