Anti-SRM Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB111884-50EA 560 EUR
ABCAAB111884-50
Anti-SRM Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SRM (spermidine synthase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR