Anti-KATNA1 Rabbit Monoclonal Antibody [clone: EPR5071]
Proveedor: Abcam
ABCAAB111881-100EA 490 EUR
ABCAAB111881-100
Anti-KATNA1 Rabbit Monoclonal Antibody [clone: EPR5071]
Anticuerpos
Type: Primary
Antigen: KATNA1 (katanin p60 (ATPase containing) subunit A 1)
Clonality: monoclonal
Clone: EPR5071
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now