Anti-ALK Rabbit Polyclonal Antibody
Proveedor: Abcam
ABCAAB111865-100EA 753 EUR
ABCAAB111865-100
Anti-ALK Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: ALK (anaplastic lymphoma receptor tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human