Anti-MLLT1 Rabbit Polyclonal Antibody
Proveedor: Abcam
ABCAAB111864-100EA 819 EUR
ABCAAB111864-100
Anti-MLLT1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: MLLT1 (myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human