Anti-SYNGR2 Rabbit Polyclonal Antibody
Proveedor: Abcam
ABCAAB106459-100EA 560 EUR
ABCAAB106459-100
Anti-SYNGR2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SYNGR2 (synaptogyrin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat