Anti-IgG Goat Polyclonal Antibody (Biotin)
Proveedor: Abcam
ABCAAB98395-500EA 0 EUR
ABCAAB98395-500
Anti-IgG Goat Polyclonal Antibody (Biotin)
Anticuerpos
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now