Anti-Cytochrome C oxidase subunit II Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB91317-100EA 575 EUR
ABCAAB91317-100
Anti-Cytochrome C oxidase subunit II Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Cytochrome C oxidase subunit II
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR