Anti-MPO Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 2D4]
Proveedor: Abcam
ABCAAB90812-50EA 550 EUR
ABCAAB90812-50
Anti-MPO Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 2D4]
Anticuerpos
Type: Primary
Antigen: MPO (myeloperoxidase)
Clonality: monoclonal
Clone: 2D4
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat