Anti-l Afadin Mouse Monoclonal Antibody [clone: 3]

Proveedor: Abcam
ABCAAB90809-50EA 739 EUR
ABCAAB90809-50
Anti-l Afadin Mouse Monoclonal Antibody [clone: 3]
Anticuerpos
Type: Primary
Antigen: l Afadin
Clonality: monoclonal
Clone: 3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR