Anti-Sarcomeric Alpha Actinin Rabbit Polyclonal Antibody
Proveedor: Abcam
ABCAAB90776-100EA 555 EUR
ABCAAB90776-100
Anti-Sarcomeric Alpha Actinin Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Sarcomeric Alpha Actinin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human