Anti-GRM1 Rabbit polyclonal antibody unconjugated

ABCAAB90756-100EA 713 EUR
ABCAAB90756-100
Anti-GRM1 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: GRM1 (glutamate receptor, metabotropic 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR