Anti-ENPP3 Mouse monoclonal antibody unconjugated [clone: NP4D6]
Proveedor: Abcam
ABCAAB90754-100EA 495 EUR
ABCAAB90754-100
Anti-ENPP3 Mouse monoclonal antibody unconjugated [clone: NP4D6]
Anticuerpos
Type: Primary
Antigen: ENPP3 (ectonucleotide pyrophosphatase/phosphodiesterase 3)
Clonality: monoclonal
Clone: NP4D6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human