Anti-ENPP3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: NP4D6]
Proveedor: Abcam
ABCAAB90751-50EA 275 EUR
ABCAAB90751-50
Anti-ENPP3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: NP4D6]
Anticuerpos
Type: Primary
Antigen: ENPP3 (ectonucleotide pyrophosphatase/phosphodiesterase 3)
Clonality: monoclonal
Clone: NP4D6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human