Anti-6X His tag Rabbit Polyclonal Antibody (Agarose)
Proveedor: Abcam
ABCAAB90663-100EA 550 EUR
ABCAAB90663-100
Anti-6X His tag Rabbit Polyclonal Antibody (Agarose)
Anticuerpos
Type: Primary
Antigen: 6X His tag (6X His tag)
Clonality: polyclonal
Clone:
Conjugation: Agarose
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now