Anti-EMR1 Rat monoclonal antibody unconjugated [clone: F4/80]

Proveedor: Abcam
ABCAAB90247-100EA 485 EUR
ABCAAB90247-100
Anti-EMR1 Rat monoclonal antibody unconjugated [clone: F4/80]
Anticuerpos
Type: Primary
Antigen: EMR1 (egf-like module containing, mucin-like, hormone receptor-like 1)
Clonality: monoclonal
Clone: F4/80
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR