Anti-ULBP3 Mouse Monoclonal Antibody [clone: MM0594-6E12]
Proveedor: Abcam
ABCAAB89931-50EA 525 EUR
ABCAAB89931-50
Anti-ULBP3 Mouse Monoclonal Antibody [clone: MM0594-6E12]
Anticuerpos
Type: Primary
Antigen: ULBP3 (UL16 binding protein 3)
Clonality: monoclonal
Clone: MM0594-6E12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human