Anti-TACD2 Mouse Monoclonal Antibody [clone: MM0588-49D6]
Proveedor: Abcam
ABCAAB89928-50EA 0 EUR
ABCAAB89928-50
Anti-TACD2 Mouse Monoclonal Antibody [clone: MM0588-49D6]
Anticuerpos
Type: Primary
Antigen: TACD2
Clonality: monoclonal
Clone: MM0588-49D6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human