Anti-TNFRSF11A Mouse monoclonal antibody unconjugated [clone: MM0521-7G22]
ABCAAB89912-50EA 720 EUR
ABCAAB89912-50
Anti-TNFRSF11A Mouse monoclonal antibody unconjugated [clone: MM0521-7G22]
Type: Primary
Antigen: TNFRSF11A (tumor necrosis factor receptor superfamily, member 11a, NFKB activator)
Clonality: monoclonal
Clone: MM0521-7G22
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human