Anti-MAP3K12 Mouse Monoclonal Antibody [clone: MM0514-9D8]
Proveedor: Abcam
ABCAAB89908-50EA 480 EUR
ABCAAB89908-50
Anti-MAP3K12 Mouse Monoclonal Antibody [clone: MM0514-9D8]
Anticuerpos
Type: Primary
Antigen: MAP3K12 (mitogen-activated protein kinase kinase kinase 12)
Clonality: monoclonal
Clone: MM0514-9D8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human