Anti-UBE2V1 Rabbit Polyclonal Antibody
Proveedor: Abcam
ABCAAB89903-50EA 746 EUR
ABCAAB89903-50
Anti-UBE2V1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: UBE2V1 (ubiquitin-conjugating enzyme E2 variant 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human