Anti-PAPPA2 Mouse monoclonal antibody [clone: MM0507-8M12]

ABCAAB89899-100EA 624 EUR
ABCAAB89899-100
Anti-PAPPA2 Mouse monoclonal antibody [clone: MM0507-8M12]
Type: Primary
Antigen: PAPPA2 (pappalysin 2)
Clonality: monoclonal
Clone: MM0507-8M12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR