Anti-TNFRSF4 Mouse Monoclonal Antibody [clone: MM0505-8S23]
Proveedor: Abcam
ABCAAB89896-100EA 776 EUR
ABCAAB89896-100
Anti-TNFRSF4 Mouse Monoclonal Antibody [clone: MM0505-8S23]
Anticuerpos
Type: Primary
Antigen: TNFRSF4 (tumor necrosis factor receptor superfamily, member 4)
Clonality: monoclonal
Clone: MM0505-8S23
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human