Anti-IL15RA Mouse Monoclonal Antibody [clone: MM0369-5M53]

Proveedor: Abcam
ABCAAB89894-100EA 739 EUR
ABCAAB89894-100
Anti-IL15RA Mouse Monoclonal Antibody [clone: MM0369-5M53]
Anticuerpos
Type: Primary
Antigen: IL15RA (interleukin 15 receptor, alpha)
Clonality: monoclonal
Clone: MM0369-5M53
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR