Anti-IL12RB1 Mouse Monoclonal Antibody [clone: MM0363-7V24]
Proveedor: Abcam
ABCAAB89890-100EA 739 EUR
ABCAAB89890-100
Anti-IL12RB1 Mouse Monoclonal Antibody [clone: MM0363-7V24]
Anticuerpos
Type: Primary
Antigen: IL12RB1 (interleukin 12 receptor, beta 1)
Clonality: monoclonal
Clone: MM0363-7V24
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human