Anti-Orexin B Mouse Monoclonal Antibody [clone: MM0501-5G21]
Proveedor: Abcam
ABCAAB89888-100EA 510 EUR
ABCAAB89888-100
Anti-Orexin B Mouse Monoclonal Antibody [clone: MM0501-5G21]
Anticuerpos
Type: Primary
Antigen: Orexin B
Clonality: monoclonal
Clone: MM0501-5G21
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human