Anti-Nogo Receptor Mouse monoclonal antibody unconjugated [clone: MM0493-6H43]
ABCAAB89879-100EA 696 EUR
ABCAAB89879-100
Anti-Nogo Receptor Mouse monoclonal antibody unconjugated [clone: MM0493-6H43]
Type: Primary
Antigen: Nogo Receptor
Clonality: monoclonal
Clone: MM0493-6H43
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human