Anti-NCR1 Mouse Monoclonal Antibody [clone: MM0491-8F24]
Proveedor: Abcam
ABCAAB89877-50EA 550 EUR
ABCAAB89877-50
Anti-NCR1 Mouse Monoclonal Antibody [clone: MM0491-8F24]
Anticuerpos
Type: Primary
Antigen: NCR1 (natural cytotoxicity triggering receptor 1)
Clonality: monoclonal
Clone: MM0491-8F24
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human