Anti-non-muscle Myosin IIA Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB89837-100EA 545 EUR
ABCAAB89837-100
Anti-non-muscle Myosin IIA Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: non-muscle Myosin IIA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR