Anti-LECT2 Mouse Monoclonal Antibody [clone: MM0449-4B24]
Proveedor: Abcam
ABCAAB89835-50EA 525 EUR
ABCAAB89835-50
Anti-LECT2 Mouse Monoclonal Antibody [clone: MM0449-4B24]
Anticuerpos
Type: Primary
Antigen: LECT2 (leukocyte cell-derived chemotaxin 2)
Clonality: monoclonal
Clone: MM0449-4B24
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human