Anti-DUSP12 Mouse Polyclonal Antibody
Proveedor: Abcam
ABCAAB89827-50EA 712 EUR
ABCAAB89827-50
Anti-DUSP12 Mouse Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: DUSP12 (dual specificity phosphatase 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human