Anti-OPTN Mouse polyclonal antibody unconjugated
ABCAAB89820-50EA 720 EUR
ABCAAB89820-50
Anti-OPTN Mouse polyclonal antibody unconjugated
Type: Primary
Antigen: OPTN (optineurin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human