Anti-LCN2 Mouse Polyclonal Antibody

Proveedor: Abcam
ABCAAB89819-50EA 560 EUR
ABCAAB89819-50
Anti-LCN2 Mouse Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: LCN2 (lipocalin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR