Anti-HCN2 Mouse monoclonal antibody [clone: S71-37]

Proveedor: Abcam
ABCAAB84817-100EA 495 EUR
ABCAAB84817-100
Anti-HCN2 Mouse monoclonal antibody [clone: S71-37]
Type: Primary
Antigen: HCN2 (hyperpolarization activated cyclic nucleotide-gated potassium channel 2)
Clonality: monoclonal
Clone: S71-37
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR